1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
CAS: 1633-83-6 | C4H8O3S | 136.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR107675000
Price (MYR)
3,680.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,680.00
Add to cart
Specifications
Chemical Name or Material1, 4-Butane sultone
Name Note99+%
CAS1633-83-6
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning