2,3-Butanedione, 99%, Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
CAS: 431-03-8 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR107655000
Price (MYR)
1,176.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,176.00
Add to cart
Chemical Identifiers
CAS13268-42-3
IUPAC Nametriammonium hexaoxo-1,4,6,9,10,13-hexaoxa-5-ferraspiro[4.4⁵.4⁵]tridecane-5,5,5-tris(ylium) trihydrate
Molecular FormulaC6H18FeN3O15
InChI KeyARJSTCBPHIPXIJ-UHFFFAOYSA-K
SMILES[NH4+].[NH4+].[NH4+].O.O.O.O=C1O[Fe+3]234(OC1=O)OC(=O)C(=O)O2.O=C(O3)C(=O)O4
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
Appearance (Form)or fused solid
Titration with KMnO497.5 to 102.5 %
Appearance (Form)Adhering crystals or crystalline powder