2,3-Butanedione, 99%, Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
CAS: 431-03-8 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR107651000
Price (MYR)
759.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
759.00
Add to cart
Chemical Identifiers
CAS814-87-9
IUPAC Namedialuminium(3+) trioxalate
Molecular FormulaC6Al2O12
InChI KeyZCLVNIZJEKLGFA-UHFFFAOYSA-H
SMILES[Al+3].[Al+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or chunks
Assay (unspecified)≥98.0%
Water ContentReport waters of hydration