Butadiene sulfone, 98%, Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
CAS: 77-79-2 | C4H6O2S | 118.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR107615000
Price (MYR)
803.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
803.00
Add to cart
Specifications
Chemical Name or MaterialButadiene sulfone
CAS77-79-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
Health Hazard 3GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/eye protec