N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR107451000
Price (MYR)
511.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
511.00
Add to cart
Chemical Identifiers
CAS7783-56-4
IUPAC Nameantimony(3+) trifluoride
Molecular FormulaF3Sb
InChI KeyGUNJVIDCYZYFGV-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Sb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodimetric>=97.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White to grey
Coupling reagent, C-X bond formation reagent
RUO – Research Use Only