N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR107450100
Price (MYR)
514.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
514.00
Add to cart
Chemical Identifiers
CAS108-26-9
IUPAC Name3-methyl-4,5-dihydro-1H-pyrazol-5-one
Molecular FormulaC4H6N2O
InChI KeyNHLAPJMCARJFOG-UHFFFAOYSA-N
SMILESCC1=NNC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to cream to pale yellow
Coupling reagent, C-X bond formation reagent
RUO – Research Use Only