N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
10 g
100 g
1 kg
Packaging:
View all
Plastic drum
Catalog number ACR107450050
Price (MYR)
4,365.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
4,365.00
Add to cart
Specifications
Chemical Name or MaterialN-Bromosuccinimide
CAS128-08-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Coupling reagent, C-X bond formation reagent
RUO – Research Use Only