4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromopyridine Hydrochloride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number ACR107420050
Price (MYR)
761.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
761.00
Add to cart
Specifications
Chemical Name or Material4-Bromopyridine hydrochloride
CAS19524-06-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0