3-Bromopyridine, 99%, Thermo Scientific Chemicals
3-Bromopyridine, 99%, Thermo Scientific Chemicals
3-Bromopyridine, 99%, Thermo Scientific Chemicals
3-Bromopyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromopyridine, 99%, Thermo Scientific Chemicals
CAS: 626-55-1 | C5H4BrN | 158.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Packaging:
Glass bottle
Catalog number ACR107411000
Price (MYR)
1,488.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,488.00
Add to cart
Chemical Identifiers
CAS149104-90-5
IUPAC Name(4-acetylphenyl)boronic acid
Molecular FormulaC8H9BO3
InChI KeyOBQRODBYVNIZJU-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow to pale brown
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5%
Assay (HPLC)≥97.5%
Proton NMRConforms to structure