3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
CAS: 5003-71-4 | C3H8BrN·HBr | 218.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR107381000
Price (MYR)
1,183.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,183.00
Add to cart
Specifications
Chemical Name or Material3-Bromopropylamine hydrobromide
Name Note98%
CAS5003-71-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning