3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals
CAS: 5003-71-4 | C3H8BrN·HBr | 218.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR107380250
Price (MYR)
575.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
575.00
Add to cart
Chemical Identifiers
CAS6001-64-5
IUPAC Namebis(1,1,1-trichloro-2-methylpropan-2-ol) hydrate
Molecular FormulaC8H16Cl6O3
InChI KeyWRWLCXJYIMRJIN-UHFFFAOYSA-N
SMILESO.CC(C)(O)C(Cl)(Cl)Cl.CC(C)(O)C(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 % (ex Cl)
Appearance (Color)White to off-white
Water4.5 to 5.5 % (K.F.)