1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
CAS: 637-59-2 | C9H11Br | 199.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR107295000
Price (MYR)
3,499.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,499.00
Add to cart
Chemical Identifiers
CAS2623-91-8
IUPAC Name(2R)-2-aminobutanoic acid
Molecular FormulaC4H9NO2
InChI KeyQWCKQJZIFLGMSD-GSVOUGTGSA-N
SMILESCC[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Optical Rotation-21.0 ± 1.0? (C=5 in 6N HCl)
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%