1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
CAS: 637-59-2 | C9H11Br | 199.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR107291000
Price (MYR)
1,107.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,107.00
Add to cart
Specifications
Chemical Name or Material1-Bromo-3-phenylpropane
Name Note98%
CAS637-59-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning