1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
CAS: 637-59-2 | C9H11Br | 199.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR107290050
Price (MYR)
365.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
365.00
Add to cart
Chemical Identifiers
CAS345-16-4
IUPAC Name5-fluoro-2-hydroxybenzoic acid
Molecular FormulaC7H5FO3
InChI KeyJWPRICQKUNODPZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(F)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥96.0%
Appearance (Color)White
Melting Point (clear melt)175.0-185.0?C
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%