1-Bromonaphthalene, 96%, Thermo Scientific Chemicals
1-Bromonaphthalene, 96%, Thermo Scientific Chemicals
1-Bromonaphthalene, 96%, Thermo Scientific Chemicals
1-Bromonaphthalene, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromonaphthalene, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR107155000
Price (MYR)
2,270.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,270.00
Add to cart
Specifications
Chemical Name or Material1-Bromonaphthalene
Name Note96%
CAS90-11-9
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning