5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
CAS: 10075-50-0 | C8H6BrN | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number ACR107060250
Price (MYR)
823.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
823.00
Add to cart
Specifications
Chemical Name or Material5-Bromoindole
Name Note99%
CAS10075-50-0
CAS Min %98.5
CAS Max %100.0