5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
CAS: 10075-50-0 | C8H6BrN | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR107060050
Price (MYR)
410.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
410.00
Add to cart
Chemical Identifiers
CAS6032-29-7
IUPAC Namepentan-2-ol
Molecular FormulaC5H12O
InChI KeyJYVLIDXNZAXMDK-UHFFFAOYNA-N
SMILESCCCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4050-1.4080 @ 20?C