4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 460-00-4 | C6H4BrF | 175.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass bottle
Catalog number ACR107010100
Price (MYR)
406.00
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
406.00
Add to cart
Chemical Identifiers
CAS471-47-6
IUPAC Namecarbamoylformic acid
Molecular FormulaC2H3NO3
InChI KeySOWBFZRMHSNYGE-UHFFFAOYSA-N
SMILESNC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (special)≥97.5 to ≤102.5% (based on elemetal H, C, N)