3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Catalog number ACR107000250
Price (MYR)
531.00
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (MYR)
531.00
Add to cart
Chemical Identifiers
CAS705-76-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)44.5-50.5?C