2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 1072-85-1 | C6H4BrF | 175 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass bottle
Catalog number ACR106990100
Price (MYR)
530.00
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
530.00
Add to cart
Chemical Identifiers
CAS623-48-3
IUPAC Nameethyl 2-iodoacetate
Molecular FormulaC4H7IO2
InChI KeyMFFXVVHUKRKXCI-UHFFFAOYSA-N
SMILESCCOC(=O)CI
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
StabilizerCopper wire
Appearance (Color)Clear colorless or pale yellow to orange
Assay (GC)≥97.0%
Refractive Index1.5020-1.5070 @ 20?C