2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
CAS: 2576-47-8 | C2H6BrN·HBr | 204.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR106935000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1788-31-4
IUPAC NameN-(1,3-diphenylpropan-2-ylidene)hydroxylamine
Molecular FormulaC15H15NO
InChI KeySXEBLVKLMOIGER-UHFFFAOYSA-N
SMILESON=C(CC1=CC=CC=C1)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.0%
Melting Point (clear melt)119.5-125.5?C