2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
CAS: 2576-47-8 | C2H6BrN·HBr | 204.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
250 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number ACR106931000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS71-91-0
IUPAC Nametetraethylazanium bromide
Molecular FormulaC8H20BrN
InChI KeyHWCKGOZZJDHMNC-UHFFFAOYSA-M
SMILES[Br-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration Argentometric>=99.0 %
Appearance (Color)White to cream
Loss on drying=<1.0 %