4-Bromobenzonitrile, 98%, Thermo Scientific Chemicals
4-Bromobenzonitrile, 98%, Thermo Scientific Chemicals
4-Bromobenzonitrile, 98%, Thermo Scientific Chemicals
4-Bromobenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromobenzonitrile, 98%, Thermo Scientific Chemicals
CAS: 623-00-7 | C7H4BrN | 182.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass Bottle
Catalog number ACR106710100
Price (MYR)
466.00
Add to cart
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
466.00
Add to cart
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.