4-Bromoanisole, 98%, Thermo Scientific Chemicals
4-Bromoanisole, 98%, Thermo Scientific Chemicals
4-Bromoanisole, 98%, Thermo Scientific Chemicals
4-Bromoanisole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromoanisole, 98%, Thermo Scientific Chemicals
CAS: 104-92-7 | C7H7BrO | 187.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Catalog number ACR106631000
Price (MYR)
583.00
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
583.00
Add to cart
Chemical Identifiers
CAS25952-53-8
IUPAC Name({[3-(dimethylamino)propyl]imino}methylidene)(ethyl)amine hydrochloride
Molecular FormulaC8H18ClN3
InChI KeyFPQQSJJWHUJYPU-UHFFFAOYSA-N
SMILESCl.CCN=C=NCCCN(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point111°C to 116°C
Titration Argentometric>=98.0 %