Biuret, 97%, extra pure
Biuret, 97%, extra pure
Biuret, 97%, extra pure
Biuret, 97%, extra pure
Thermo Scientific Chemicals
Biuret, 97%, extra pure
CAS: 108-19-0 | C2H5N3O2 | 103.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR106521000
Price (MYR)
1,467.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,467.00
Add to cart
Chemical Identifiers
CAS7782-85-6
IUPAC Namedisodium phosphoric acid heptahydrate
Molecular FormulaH17Na2O11P
InChI KeyPYLIXCKOHOHGKQ-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<5 ppm
Arsenic (As)=<5 ppm
Sulfate (SO4)=<50 ppm
Potassium (K)=<100 ppm
Appearance (Color)White