Methylthymol blue, tetrasodium salt, pure, water soluble
Methylthymol blue, tetrasodium salt, pure, water soluble
Methylthymol blue, tetrasodium salt, pure, water soluble
Methylthymol blue, tetrasodium salt, pure, water soluble
Thermo Scientific Chemicals
Methylthymol blue, tetrasodium salt, pure, water soluble
CAS: 1945-77-3 | C37H44N2Na4O13S | 848.779 g/mol
Have Questions?
Catalog NumberQuantity
ACR1063700505 g
Catalog number ACR106370050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS872-50-4
IUPAC Name1-methylpyrrolidin-2-one
Molecular FormulaC5H9NO
InChI KeySECXISVLQFMRJM-UHFFFAOYSA-N
SMILESCN1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Water=<0.05 % (K.F.)
Infrared spectrumConforms
Specific gravity(20°C) 1.02 to 1.04
View more
Indicator
RUO – Research Use Only