Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number ACR106230250
Price (MYR)
458.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
458.00
Add to cart
Chemical Identifiers
CAS141-78-6
IUPAC Nameethyl acetate
Molecular FormulaC4H8O2
InChI KeyXEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILESCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Water=<0.05 % (K.F.)
Residue after evaporation=<0.005 %
Appearance (Form)Clear liquid
Color scale=<10 APHA