Benzylamine, 99%, pure, Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
CAS: 100-46-9 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number ACR105855000
Price (MYR)
647.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
647.00
Add to cart
Chemical Identifiers
CAS501-52-0
IUPAC Name3-phenylpropanoic acid
Molecular FormulaC9H10O2
InChI KeyXMIIGOLPHOKFCH-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or mass and/or
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White to yellow to light beige
Appearance (Form)chunks