Benzylamine, 99%, pure, Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
CAS: 100-46-9 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR105850025
Price (MYR)
1,634.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,634.00
Add to cart
Chemical Identifiers
CAS331-39-5
IUPAC Name(2Z)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
Molecular FormulaC9H8O4
InChI KeyQAIPRVGONGVQAS-RQOWECAXSA-N
SMILESOC(=O)C=C/C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with NaOH>=99.0 %
Appearance (Color)Yellow to yellow-green