Benzoylhydrazine, 98%, Thermo Scientific Chemicals
Benzoylhydrazine, 98%, Thermo Scientific Chemicals
Benzoylhydrazine, 98%, Thermo Scientific Chemicals
Benzoylhydrazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoylhydrazine, 98%, Thermo Scientific Chemicals
CAS: 613-94-5 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR105775000
Price (MYR)
2,067.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
2,067.00
Add to cart
Chemical Identifiers
CAS13494-80-9
IUPAC Nametellurium
Molecular FormulaTe
InChI KeyPORWMNRCUJJQNO-UHFFFAOYSA-N
SMILES[Te]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.