Benzoylhydrazine, 98%, Thermo Scientific Chemicals
Benzoylhydrazine, 98%, Thermo Scientific Chemicals
Benzoylhydrazine, 98%, Thermo Scientific Chemicals
Benzoylhydrazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoylhydrazine, 98%, Thermo Scientific Chemicals
CAS: 613-94-5 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR105771000
Price (MYR)
646.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
646.00
Add to cart
Chemical Identifiers
CAS402-24-4
IUPAC Name1-ethenyl-3-(trifluoromethyl)benzene
Molecular FormulaC9H7F3
InChI KeyARHOUOIHKWELMD-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(C=C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4640 to 1.4660 (20°C, 589 nm)
StabilizerAbout 100 ppm 4-tert-Butylcatechol
Appearance (Form)Liquid
GC>=97.0 %
Appearance (Color)Clear colorless