2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Benzoylbenzoic Acid, 98+%, Thermo Scientific Chemicals
CAS: 85-52-9 | C14H10O3 | 226.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Plastic bottle
Catalog number ACR105710250
Price (MYR)
375.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
375.00
Add to cart
Specifications
Chemical Name or Material2-Benzoylbenzoic acid
Name Note98+%
CAS85-52-9
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning