Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
5 mg
25 mg
Packaging:
Glass bottle
Catalog number ACR105551000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10349-57-2
IUPAC Namequinoline-6-carboxylic acid
Molecular FormulaC10H7NO2
InChI KeyVXGYRCVTBHVXMZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2N=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Water=<2 %
Appearance (Color)Beige to brown