alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals
alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals
alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals
alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals
CAS: 441-38-3 | C14H13NO2 | 227.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR105530250
Price (MYR)
506.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
506.00
Add to cart
Chemical Identifiers
CAS98330-04-2
IUPAC Nametetraethylazanium fluoride
Molecular FormulaC8H20FN
InChI KeyQSUJAUYJBJRLKV-UHFFFAOYSA-M
SMILES[F-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)< 28% (non-U.S. specification)
Assay (Non-aqueous acid-base Titration)≥96.0% (dry wt. basis, non-U.S. specification)
FormPowder or crystals or crystalline powder
Appearance (Color)White to pale cream
Assay from Supplier's CofA≥96.0% (U.S. specification)