Benzoin, 98%, Thermo Scientific Chemicals
Benzoin, 98%, Thermo Scientific Chemicals
Benzoin, 98%, Thermo Scientific Chemicals
Benzoin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoin, 98%, Thermo Scientific Chemicals
CAS: 119-53-9 | C14H12O2 | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR105511000
Price (MYR)
355.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
355.00
Add to cart
Chemical Identifiers
CAS1429056-28-9
IUPAC Namebis(2-oxa-7-azaspiro[3.5]nonane); oxalic acid
Molecular FormulaC16H28N2O6
InChI KeyWWVUFRRXXSVWBJ-UHFFFAOYSA-N
SMILESOC(=O)C(O)=O.C1OCC11CCNCC1.C1OCC11CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)96.0% min