Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
CAS: 91-01-0 | C13H12O | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR105395000
Price (MYR)
931.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
931.00
Add to cart
Specifications
Chemical Name or MaterialBenzhydrol
Name Note99%
CAS91-01-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning