Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzhydrol, 99%, Thermo Scientific Chemicals
CAS: 91-01-0 | C13H12O | 184.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR105391000
Price (MYR)
438.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
438.00
Add to cart
Chemical Identifiers
CAS7789-06-2
IUPAC Namestrontium(2+) dioxochromiumbis(olate)
Molecular FormulaCrO4Sr
InChI KeyNVKTUNLPFJHLCG-UHFFFAOYSA-N
SMILES[Sr++].[O-][Cr]([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95.0% min