1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
CAS: 89-32-7 | C10H2O6 | 218.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number ACR105325000
Price (MYR)
1,114.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,114.00
Add to cart
Chemical Identifiers
CAS124-09-4
IUPAC Namehexane-1,6-diamine
Molecular FormulaC6H16N2
InChI KeyNAQMVNRVTILPCV-UHFFFAOYSA-N
SMILESNCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or fused solid
Assay (GC)≥98.0%
CommentMay discolor on storage
Water Content (Karl Fischer Titration)<1.0%