1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic anhydride, 99%, Thermo Scientific Chemicals
CAS: 89-32-7 | C10H2O6 | 218.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR105321000
Price (MYR)
467.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
467.00
Add to cart
Chemical Identifiers
CAS14726-36-4
IUPAC Namezinc(2+) bis((dibenzylcarbamothioyl)sulfanide)
Molecular FormulaC30H28N2S4Zn
InChI KeyAUMBZPPBWALQRO-UHFFFAOYSA-L
SMILES[Zn++].[S-]C(=S)N(CC1=CC=CC=C1)CC1=CC=CC=C1.[S-]C(=S)N(CC1=CC=CC=C1)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Melting point183°C to 188°C
Assay>=94 % (ex Zn)
Appearance (Color)White to off-white