1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
View all
Plastic bottle
Catalog number ACR105311000
Price (MYR)
405.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
405.00
Add to cart
Specifications
Chemical Name or Material1,2,4,5-Benzenetetracarboxylic acid
Name Note96%
CAS89-05-4
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning