1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,2,4,5-Benzenetetracarboxylic acid, 96%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
View all
Glass bottle
Catalog number ACR105310050
Price (MYR)
345.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
345.00
Add to cart
Specifications
Chemical Name or Material1,2,4,5-Benzenetetracarboxylic acid
Name Note96%
CAS89-05-4
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning