Antipyrine, 99%, Thermo Scientific Chemicals
Antipyrine, 99%, Thermo Scientific Chemicals
Antipyrine, 99%, Thermo Scientific Chemicals
Antipyrine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Antipyrine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR104971000
Price (MYR)
438.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
438.00
Add to cart
Specifications
Chemical Name or MaterialAntipyrine
Name Note99%
CAS60-80-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement