Anthranilic acid, 98+%
Anthranilic acid, 98+%
Anthranilic acid, 98+%
Anthranilic acid, 98+%
Thermo Scientific Chemicals
Anthranilic acid, 98+%
Anthranilic acid, CAS # 118-92-3, also known as 2-Aminobenzoic acid or o-amino-benzoic acid, is an organic aromatic acid. | CAS: 118-92-3 | C7H7NO2 | 137.14 g/mol
Have Questions?
Catalog NumberQuantity
ACR104915000500 g
Catalog number ACR104915000
Price (MYR)
520.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
520.00
Add to cart
Chemical Identifiers
CAS93267-04-0
IUPAC Namemethyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-iodopropanoate
Molecular FormulaC9H16INO4
InChI KeyUGZBFCCHLUWCQI-LURJTMIESA-N
SMILESCOC(=O)[C@H](CI)NC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point49°C to 53°C
Specific optical rotation-3.0° to -5.0° (20°C, 589 nm) (c=2 in methanol)
Appearance (Color)White to yellow
Infrared spectrumConforms
View more
General Description
  • Anthranillic acid is an organic aromatic acid consisting of benzoic acid with an o-substituted amino group
  • Produced as a metabolite in the L-Tryptophan-kynurenine pathway, anthranilic acid is found in both mice and humans
  • Esters of anthranilic acid are used in perfumes, dyes, synthetic flavor development, and as bird and insect repellents
  • Anthranilic acid has been used as a fungicide/mold inhibitor
Applications
  • Anthranilic acid is an organic reagent used in preparation of dyes, pigments, and perfumes
  • It has been used to fluorescently tag reducing glycans for analysis by HPLC or LC/MS
  • It can be used as a fluorophore group in FRET applications
RUO – Research Use Only
General References:
  1. Bigge, J.C.; Patel, T.P.; Bruce, J.A.; Goulding, P.N.; Charles, S.M.; Parekh, R.B. Nonselective and efficient fluorescent labeling of glycans using 2-amino benzamide and anthranilic acid. Anal Biochem. 1995, 230(2), 229-38.
  2. Zhu, Y.; Liu, X.; Zhang, Y.; Wang, Z.; Lasanajak, Y.; Song, X. Anthranilic Acid as a Versatile Fluorescent Tag and Linker for Functional Glycomics. Bioconjug Chem. 2018, 29(11), 3847-3855.