Anthranilic acid, 98+%, Thermo Scientific Chemicals
Anthranilic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Anthranilic acid, 98+%, Thermo Scientific Chemicals
CAS: 118-92-3 | C7;H7;NO2; | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Catalog number ACR104911000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS130-85-8
IUPAC Name4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
Molecular FormulaC23H16O6
InChI KeyWLJNZVDCPSBLRP-UHFFFAOYSA-N
SMILESOC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration excess NaOH98.0 to 102.0 %
Appearance (Color)Yellow to yellow-green