Anthracene, 99%, Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
CAS: 120-12-7 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
View all
Plastic bottle
Catalog number ACR104861000
Price (MYR)
620.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
620.00
Add to cart
Chemical Identifiers
CAS2627-69-2
IUPAC Name5-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-imidazole-4-carboxamide
Molecular FormulaC9H14N4O5
InChI KeyRTRQQBHATOEIAF-UUOKFMHZSA-N
SMILESNC(=O)C1=C(N)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Appearance (Form)Crystalline powder
Infrared spectrumConforms
TLC>=97.5 %
Specific optical rotation-62.0° to -66.0° (20°C, 589 nm) (c=1 H2O)