Anthracene, 99%, Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Anthracene, 99%, Thermo Scientific Chemicals
CAS: 120-12-7 | C14H10 | 178.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
Catalog number ACR104860050
Price (MYR)
391.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
391.00
Add to cart
Chemical Identifiers
CAS894086-00-1
IUPAC Name5-(di-tert-butylphosphanyl)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole
Molecular FormulaC32H35N4P
InChI KeyPTXJGGGNGMPMBG-UHFFFAOYSA-N
SMILESCC(C)(C)P(C1=CC=NN1C1=C(N(N=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Proton NMR≥ 96.0%