p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
CAS: 104-94-9 | C7H9NO | 123.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Glass bottle
Catalog number ACR104832500
Price (MYR)
549.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
549.00
Add to cart
Chemical Identifiers
CAS556-67-2
IUPAC Nameoctamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular FormulaC8H24O4Si4
InChI KeyHMMGMWAXVFQUOA-UHFFFAOYSA-N
SMILESC[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.3950 to 1.3970 (20°C, 589 nm)