p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
p-Anisidine, 99%, Thermo Scientific Chemicals
CAS: 104-94-9 | C7H9NO | 123.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
250 g
Packaging:
Glass bottle
Catalog number ACR104830010
Price (MYR)
1,121.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,121.00
Add to cart
Chemical Identifiers
CAS493-09-4
IUPAC Name2,3-dihydro-1,4-benzodioxine
Molecular FormulaC8H8O2
InChI KeyBNBQRQQYDMDJAH-UHFFFAOYSA-N
SMILESC1COC2=CC=CC=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5475-1.5515 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%