4-Aminothiophenol, 96%, Thermo Scientific Chemicals
4-Aminothiophenol, 96%, Thermo Scientific Chemicals
4-Aminothiophenol, 96%, Thermo Scientific Chemicals
4-Aminothiophenol, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Aminothiophenol, 96%, Thermo Scientific Chemicals
CAS: 1193-02-8 | C6H6NS | 124.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number ACR104680050
Price (MYR)
518.00
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
518.00
Add to cart
Specifications
Chemical Name or Material4-Aminothiophenol
Name Note96%
CAS1193-02-8
CAS Min %95.0
Health Hazard 1GHS Signal Word: Danger