2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminopyrimidine, 98%, Thermo Scientific Chemicals
CAS: 109-12-6 | C4H5N3 | 95.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number ACR104581000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1003-99-2
IUPAC Name2-bromo-5-fluoroaniline
Molecular FormulaC6H5BrFN
InChI KeyFWTXFEKVHSFTDQ-UHFFFAOYSA-N
SMILESNC1=CC(F)=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Melting Point (clear melt)40.0-47.0°C
FormCrystals or powder or crystalline powder or lumps or fused solid
Appearance (Color)White to cream to dark brown